Methyl-3-chloro-1-methyl-2-oxo-1,2-dihydroquinoxaline-6-carboxylate ≥95%

Supplier: Matrix Scientific

058250-500MG
101914-400EA 1258.46 USD
101914-400
Methyl-3-chloro-1-methyl-2-oxo-1,2-dihydroquinoxaline-6-carboxylate ≥95%
Methyl-3-chloro-1-methyl-2-oxo-1,2-dihydroquinoxaline-6-carboxylate

Hazard: Irritant

Formula: C₁₁H₉ClN₂O₃
MDL Number: MFCD17167318
CAS Number: 1246738-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR