Methyl-2-(2H-1,3-benzodioxol-5-yl)acetate ≥95%
Supplier: Matrix Scientific
058248-500MG
058248-1G
101914-394EA
183.08
USD
101914-394
101914-392
Methyl-2-(2H-1,3-benzodioxol-5-yl)acetate ≥95%
Methyl-2-(2H-1,3-benzodioxol-5-yl)acetate
Hazard: Irritant
Formula:
C₁₀H₁₀O₄ |
MDL Number:
MFCD08753503 CAS Number: 326-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...