6-(Methylamino)hexan-1-ol ≥95%
Supplier: Matrix Scientific
058247-1G
058247-500MG
101914-390EA
892.31
USD
101914-390
101914-388
6-(Methylamino)hexan-1-ol ≥95%
6-(Methylamino)hexan-1-ol
Hazard: Irritant
Formula:
C₇H₁₇NO |
MDL Number:
MFCD00046026 CAS Number: 50347-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...