2-(3-Methoxyphenyl)acetohydrazide ≥95%

Supplier: Matrix Scientific

058244-500MG 058244-5G 058244-1G
101914-376EA 232.31 USD
101914-376 101914-374 101914-378
2-(3-Methoxyphenyl)acetohydrazide ≥95%
2-(3-Methoxyphenyl)acetohydrazide

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MW: 180.21 g/mol
MDL Number: MFCD00085106
CAS Number: 34624-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR