2-Chloro-3-ethylquinoxaline ≥95%

Supplier: Matrix Scientific

058240-500MG 058240-1G
101914-358EA 467.69 USD
101914-358 101914-356
2-Chloro-3-ethylquinoxaline ≥95%
2-Chloro-3-ethylquinoxaline

Hazard: Irritant

Formula: C₁₀H₉ClN₂
MDL Number: MFCD00052599
CAS Number: 77186-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR