9H-Fluoren-9-ylmethyl-N-(piperidin-3-ylmethyl)carbamate ≥95%
Supplier: Matrix Scientific
058235-500MG
058235-1G
058235-5G
101914-332EA
747.69
USD
101914-332
101914-336
101914-334
9H-Fluoren-9-ylmethyl-N-(piperidin-3-ylmethyl)carbamate ≥95%
9H-Fluoren-9-ylmethyl-N-(piperidin-3-ylmethyl)carbamate
Hazard: Irritant
Formula:
C₂₁H₂₄N₂O₂ MW: 336.44 g/mol |
MDL Number:
MFCD03001743 CAS Number: 672310-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...