4-[(9H-Fluoren-9-ylmethoxy)carbonyl]morpholine-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
058232-500MG
058232-1G
101914-324EA
120
USD
101914-324
101914-322
4-[(9H-Fluoren-9-ylmethoxy)carbonyl]morpholine-2-carboxylic acid ≥95%
4-[(9H-Fluoren-9-ylmethoxy)carbonyl]morpholine-2-carboxylic acid
Hazard: Irritant
Formula:
C₂₀H₁₉NO₅ |
MDL Number:
MFCD00800355 CAS Number: 312965-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...