1-(Ethylcarbamoyl)piperidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

058229-1G 058229-500MG
101914-310EA 532.31 USD
101914-310 101914-312
1-(Ethylcarbamoyl)piperidine-4-carboxylic acid ≥95%
1-(Ethylcarbamoyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₆N₂O₃
MDL Number: MFCD08444848
CAS Number: 923177-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR