3-Ethoxyquinoxaline-2-carboxylic acid ≥90%

Supplier: Matrix Scientific

058228-500MG
101914-308EA 881.54 USD
101914-308
3-Ethoxyquinoxaline-2-carboxylic acid ≥90%
3-Ethoxyquinoxaline-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₃
MDL Number: MFCD17167317
CAS Number: 159782-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR