5-Chloro-7-nitroisoquinolin-8-amine ≥95%

Supplier: Matrix Scientific

058214-500MG 058214-1G
101914-250EA 684.62 USD
101914-250 101914-248
5-Chloro-7-nitroisoquinolin-8-amine ≥95%
5-Chloro-7-nitroisoquinolin-8-amine

Hazard: Irritant

Formula: C₉H₆ClN₃O₂
MDL Number: MFCD17167319
CAS Number: 1246738-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR