1-Benzyl-5-(piperazin-1-yl)-1,2,3,4-tetrahydroquinazolin-2-one hydrochloride ≥95%

Supplier: Matrix Scientific

058202-1G 058202-500MG
101914-200EA 1400 USD
101914-200 101914-198
1-Benzyl-5-(piperazin-1-yl)-1,2,3,4-tetrahydroquinazolin-2-one hydrochloride ≥95%
1-Benzyl-5-(piperazin-1-yl)-1,2,3,4-tetrahydroquinazolin-2-one hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₃ClN₄O
MDL Number: MFCD17167313
CAS Number: 1246738-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR