3-(Aminomethyl)-N-methylbenzamide hydrochloride ≥95%
Supplier: Matrix Scientific
058199-500MG
101914-188EA
532.31
USD
101914-188
3-(Aminomethyl)-N-methylbenzamide hydrochloride ≥95%
3-(Aminomethyl)-N-methylbenzamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₃ClN₂O |
MDL Number:
MFCD09971582 CAS Number: 916211-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...