4-Aminotetrahydropyran hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Tetrahydro-2H-pyran-4-amine hydrochloride

058196-5G
101914-184EA 36.92 USD
101914-184
4-Aminotetrahydropyran hydrochloride ≥95%
4-Aminotetrahydropyran hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO
MW: 137.6079 g/mol
MDL Number: MFCD00100881
CAS Number: 33024-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR