Ethyl-5-hydroxy-8-(2-methoxyethyl)-7-oxo-2-phenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%

Supplier: Matrix Scientific

058193-1G
101914-172EA 1290.77 USD
101914-172
Ethyl-5-hydroxy-8-(2-methoxyethyl)-7-oxo-2-phenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%
Ethyl-5-hydroxy-8-(2-methoxyethyl)-7-oxo-2-phenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₁₉H₁₉N₃O₅
MDL Number: MFCD17011966
CAS Number: 76360-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR