Ethyl-8-benzyl-5-hydroxy-7-oxo-2-phenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%

Supplier: Matrix Scientific

058190-1G
101914-166EA 1290.77 USD
101914-166
Ethyl-8-benzyl-5-hydroxy-7-oxo-2-phenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%
Ethyl-8-benzyl-5-hydroxy-7-oxo-2-phenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₂₃H₁₉N₃O₄
MDL Number: MFCD17011963
CAS Number: 76377-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR