Ethyl-8-allyl-5-hydroxy-7-oxo-2-phenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%

Supplier: Matrix Scientific

058189-1G
101914-164EA 1290.77 USD
101914-164
Ethyl-8-allyl-5-hydroxy-7-oxo-2-phenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%
Ethyl-8-allyl-5-hydroxy-7-oxo-2-phenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₁₉H₁₇N₃O₄
MDL Number: MFCD17011962
CAS Number: 76360-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR