Ethyl-5-hydroxy-7-oxo-2,8-diphenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%

Supplier: Matrix Scientific

058187-1G
101914-160EA 1290.77 USD
101914-160
Ethyl-5-hydroxy-7-oxo-2,8-diphenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%
Ethyl-5-hydroxy-7-oxo-2,8-diphenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₂₂H₁₇N₃O₄
MDL Number: MFCD17011960
CAS Number: 76360-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR