Ethyl-5-hydroxy-8-methyl-7-oxo-2-phenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%

Supplier: Matrix Scientific

058185-1G
101914-156EA 1290.77 USD
101914-156
Ethyl-5-hydroxy-8-methyl-7-oxo-2-phenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%
Ethyl-5-hydroxy-8-methyl-7-oxo-2-phenyl-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₅N₃O₄
MDL Number: MFCD17011958
CAS Number: 1253791-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR