1-Methyl-7-phenyl-1H-pyrimido[4,5-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058175-1G
101914-136EA 806.15 USD
101914-136
1-Methyl-7-phenyl-1H-pyrimido[4,5-d][1,3]oxazine-2,4-dione ≥95%
1-Methyl-7-phenyl-1H-pyrimido[4,5-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₁₃H₉N₃O₃
MDL Number: MFCD17011948
CAS Number: 1253789-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR