7-Phenyl-1H-pyrimido[4,5-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058174-1G
101914-134EA 2343.08 USD
101914-134
7-Phenyl-1H-pyrimido[4,5-d][1,3]oxazine-2,4-dione ≥95%
7-Phenyl-1H-pyrimido[4,5-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₁₂H₇N₃O₃
MDL Number: MFCD17011947
CAS Number: 1253791-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR