Ethyl-8-benzyl-5-hydroxy-2-(methylthio)-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%

Supplier: Matrix Scientific

058168-1G
101914-124EA 1023.08 USD
101914-124
Ethyl-8-benzyl-5-hydroxy-2-(methylthio)-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%
Ethyl-8-benzyl-5-hydroxy-2-(methylthio)-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₁₈H₁₇N₃O₄S
MDL Number: MFCD17011941
CAS Number: 1253791-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR