Ethyl-5-hydroxy-2-(methylthio)-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%

Supplier: Matrix Scientific

058164-1G
101914-116EA 1023.08 USD
101914-116
Ethyl-5-hydroxy-2-(methylthio)-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate ≥95%
Ethyl-5-hydroxy-2-(methylthio)-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁N₃O₄S
MDL Number: MFCD17011937
CAS Number: 95898-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR