7-(Methylthio)-1-propyl-1H-pyrimido[4,5-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058159-1G
101914-106EA 644.62 USD
101914-106
7-(Methylthio)-1-propyl-1H-pyrimido[4,5-d][1,3]oxazine-2,4-dione ≥95%
7-(Methylthio)-1-propyl-1H-pyrimido[4,5-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₃S
MW: 253.28 g/mol
MDL Number: MFCD17011932
CAS Number: 1253789-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR