Ethyl-4-hydroxy-7-methyl-2-oxo-1-(prop-2-ynyl)-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

058152-1G 058152-250MG
101914-094EA 915.38 USD
101914-094 102551-516
Ethyl-4-hydroxy-7-methyl-2-oxo-1-(prop-2-ynyl)-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%
Ethyl-4-hydroxy-7-methyl-2-oxo-1-(prop-2-ynyl)-1,2-dihydro-1,8-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₄N₂O₄
MDL Number: MFCD17011926
CAS Number: 1253791-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR