Ethyl-1-allyl-4-hydroxy-7-methyl-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

058150-1G
101914-090EA 1049.23 USD
101914-090
Ethyl-1-allyl-4-hydroxy-7-methyl-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%
Ethyl-1-allyl-4-hydroxy-7-methyl-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₄
MDL Number: MFCD17011924
CAS Number: 69407-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR