Ethyl-1-benzyl-4-hydroxy-7-methyl-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

058148-1G
101914-086EA 3046.15 USD
101914-086
Ethyl-1-benzyl-4-hydroxy-7-methyl-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%
Ethyl-1-benzyl-4-hydroxy-7-methyl-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₉H₁₈N₂O₄
MDL Number: MFCD17011922
CAS Number: 76336-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR