Ethyl-4-hydroxy-7-methyl-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

058144-1G
101914-078EA 1049.23 USD
101914-078
Ethyl-4-hydroxy-7-methyl-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%
Ethyl-4-hydroxy-7-methyl-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₄
MDL Number: MFCD17011918
CAS Number: 76336-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR