1-(2-Methoxyethyl)-7-methyl-1H-pyrido-[2,3-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058143-1G
101914-076EA 644.62 USD
101914-076
1-(2-Methoxyethyl)-7-methyl-1H-pyrido-[2,3-d][1,3]oxazine-2,4-dione ≥95%
1-(2-Methoxyethyl)-7-methyl-1H-pyrido-[2,3-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₄
MDL Number: MFCD17011917
CAS Number: 1253792-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR