1-Isopentyl-7-methyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058141-1G
101914-072EA 644.62 USD
101914-072
1-Isopentyl-7-methyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%
1-Isopentyl-7-methyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₃
MDL Number: MFCD17011915
CAS Number: 1253789-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR