7-Methyl-1-propyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058139-1G
101914-068EA 661.54 USD
101914-068
7-Methyl-1-propyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%
7-Methyl-1-propyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MDL Number: MFCD17011913
CAS Number: 1253789-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR