1-Isopropyl-7-methyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058136-1G
101914-062EA 661.54 USD
101914-062
1-Isopropyl-7-methyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%
1-Isopropyl-7-methyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MDL Number: MFCD17011910
CAS Number: 1253791-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR