1,7-Dimethyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058135-1G
101914-060EA 661.54 USD
101914-060
1,7-Dimethyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%
1,7-Dimethyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₉H₈N₂O₃
MDL Number: MFCD17011909
CAS Number: 66690-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR