7-Methyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058134-1G
101914-058EA 661.54 USD
101914-058
7-Methyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%
7-Methyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₈H₆N₂O₃
MDL Number: MFCD17011908
CAS Number: 187543-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR