Ethyl-4-hydroxy-1-(2-methoxyethyl)-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

058133-1G
101914-056EA 903.08 USD
101914-056
Ethyl-4-hydroxy-1-(2-methoxyethyl)-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%
Ethyl-4-hydroxy-1-(2-methoxyethyl)-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₅
MDL Number: MFCD17011907
CAS Number: 1253790-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR