Ethyl-1-allyl-4-hydroxy-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

058130-1G
101914-050EA 903.08 USD
101914-050
Ethyl-1-allyl-4-hydroxy-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%
Ethyl-1-allyl-4-hydroxy-2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₄
MDL Number: MFCD17011904
CAS Number: 82360-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR