1-(2-Methoxyethyl)-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058123-1G
101914-036EA 587.69 USD
101914-036
1-(2-Methoxyethyl)-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%
1-(2-Methoxyethyl)-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₄
MDL Number: MFCD17011898
CAS Number: 1253792-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR