1-(Prop-2-ynyl)-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058122-1G 058122-250MG
101914-034EA 547.69 USD
101914-034 102551-514
1-(Prop-2-ynyl)-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%
1-(Prop-2-ynyl)-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₁₀H₆N₂O₃
MDL Number: MFCD17011897
CAS Number: 97484-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR