1-Isopentyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058121-1G
101914-032EA 587.69 USD
101914-032
1-Isopentyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%
1-Isopentyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₃
MDL Number: MFCD17011896
CAS Number: 565448-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR