1-Propyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058119-1G
101914-028EA 587.69 USD
101914-028
1-Propyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%
1-Propyl-1H-pyrido[2,3-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₃
MDL Number: MFCD17011894
CAS Number: 111396-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR