1H-Pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%

Supplier: Matrix Scientific

058114-1G
101914-018EA 592.31 USD
101914-018
1H-Pyrido[2,3-d][1,3]oxazine-2,4-dione ≥95%
1H-Pyrido[2,3-d][1,3]oxazine-2,4-dione

Hazard: Irritant

Formula: C₇H₄N₂O₃
MW: 164.12 g/mol
MDL Number: MFCD09863994
CAS Number: 21038-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR