(2S,3S,4S,5R,6R)-2,3,4,5-Tetrakis(benzyloxy)-6-bromotetrahydro-2H-pyran ≥95%

Supplier: Matrix Scientific

058079-1G
101913-996EA 515.38 USD
101913-996
(2S,3S,4S,5R,6R)-2,3,4,5-Tetrakis(benzyloxy)-6-bromotetrahydro-2H-pyran ≥95%
(2S,3S,4S,5R,6R)-2,3,4,5-Tetrakis(benzyloxy)-6-bromotetrahydro-2H-pyran

Hazard: Irritant

Formula: C₃₃H₃₃BrO₅
MDL Number: MFCD17015037
CAS Number: 1354486-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR