3,4-Dihydroxy-5-methylfuran-2(5H)-one ≥95%

Supplier: Matrix Scientific

058076-1G
101913-990EA 946.15 USD
101913-990
3,4-Dihydroxy-5-methylfuran-2(5H)-one ≥95%
3,4-Dihydroxy-5-methylfuran-2(5H)-one

Hazard: Irritant

Formula: C₅H₆O₄
MDL Number: MFCD09954510
CAS Number: 3566-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR