3-Chloro-5-(isobutylthio)-1,2,4-thiadiazole ≥95%

Supplier: Matrix Scientific

058040-1G
101913-952EA 612.31 USD
101913-952
3-Chloro-5-(isobutylthio)-1,2,4-thiadiazole ≥95%
3-Chloro-5-(isobutylthio)-1,2,4-thiadiazole

Hazard: Irritant

Formula: C₆H₉ClN₂S₂
MDL Number: MFCD15072126
CAS Number: 36955-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR