5-(Benzylthio)-3-chloro-1,2,4-thiadiazole ≥95%

Supplier: Matrix Scientific

058038-1G
101913-948EA 641.54 USD
101913-948
5-(Benzylthio)-3-chloro-1,2,4-thiadiazole ≥95%
5-(Benzylthio)-3-chloro-1,2,4-thiadiazole

Hazard: Irritant

Formula: C₉H₇ClN₂S₂
MDL Number: MFCD02948844
CAS Number: 36598-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR