5-((Cyclohexyl(methyl)amino)methyl)-1-methyl-1H-pyrazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

058032-1G
101913-936EA 1050.77 USD
101913-936
5-((Cyclohexyl(methyl)amino)methyl)-1-methyl-1H-pyrazole-3-carboxylic acid ≥95%
5-((Cyclohexyl(methyl)amino)methyl)-1-methyl-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₂₁N₃O₂
MDL Number: MFCD14708258
CAS Number: 1223748-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR