(1R,3R,4S)-Ethyl-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylate ≥95%

Supplier: Matrix Scientific

058020-1G
101913-914EA 1535.38 USD
101913-914
(1R,3R,4S)-Ethyl-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylate ≥95%
(1R,3R,4S)-Ethyl-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₅NO₂
MDL Number: MFCD14707650
CAS Number: 105786-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR