2-(Piperidin-4-yl)-5-(pyrazin-2-yl)-1,3,4-oxadiazole ≥95%

Supplier: Matrix Scientific

058010-1G
101913-894EA 2326.15 USD
101913-894
2-(Piperidin-4-yl)-5-(pyrazin-2-yl)-1,3,4-oxadiazole ≥95%
2-(Piperidin-4-yl)-5-(pyrazin-2-yl)-1,3,4-oxadiazole

Hazard: Irritant

Formula: C₁₁H₁₃N₅O
MDL Number: MFCD11846141
CAS Number: 1207175-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR