(5-Benzyl-1,3,4-oxadiazol-2-yl)methanamine ≥95%

Supplier: Matrix Scientific

058004-1G
101913-882EA 1126.15 USD
101913-882
(5-Benzyl-1,3,4-oxadiazol-2-yl)methanamine ≥95%
(5-Benzyl-1,3,4-oxadiazol-2-yl)methanamine

Hazard: Irritant

Formula: C₁₀H₁₁N₃O
MDL Number: MFCD09883724
CAS Number: 933756-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR