(5-(Pyridin-3-yl)-1,3,4-oxadiazol-2-yl)methanamine ditrifluoroacetate ≥95%

Supplier: Matrix Scientific

058000-1G
101913-874EA 1090.77 USD
101913-874
(5-(Pyridin-3-yl)-1,3,4-oxadiazol-2-yl)methanamine ditrifluoroacetate ≥95%
(5-(Pyridin-3-yl)-1,3,4-oxadiazol-2-yl)methanamine ditrifluoroacetate

Hazard: Irritant

Formula: C₁₂H₁₀F₆N₄O₅
MDL Number: MFCD28383606
CAS Number: 737690-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR