4-(3-Bromophenyl)-2-chloropyrimidine ≥95%
Supplier: Matrix Scientific
057995-1G
101913-864EA
964.62
USD
101913-864
4-(3-Bromophenyl)-2-chloropyrimidine ≥95%
4-(3-Bromophenyl)-2-chloropyrimidine
Hazard: Irritant
Formula:
C₁₀H₆BrClN₂ |
MDL Number:
MFCD11520124 CAS Number: 499195-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...